Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myo1g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134843.1 | NP_001128315.1 | 11 | 1473 | ||
XM_006251218.2 | XP_006251280.1 | 11 | 1488 | |||
XM_006251221.2 | XP_006251283.1 | 11 | 1477 | |||
XM_006251223.2 | XP_006251285.1 | 8 | 867 |