Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001115023.1 | NP_001108495.1 | 9 | 982 | ||
XM_008771088.1 | XP_008769310.1 | 9 | 1095 | |||
GenBank mRNA | BC158855.1 | - | 9 | 982 | 7930984 | |
FQ216881.1 | - | 7 | 2280 |