Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eci3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009275.1 | NP_001009275.1 | 3 | 237 | ||
GenBank mRNA | BC088178.1 | - | 3 | 237 | 7324966 | |
FQ209521.1 | - | 2 | 77 | |||
FQ223513.1 | - | 2 | 77 |