Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr141 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181767.1 | NP_861432.1 | 1 | 235 | ||
GenBank mRNA | AY288432.1 | - | 1 | 235 |
Set Membership: |
G-protein coupled receptor |