Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stard3nl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008298.1 | NP_001008299.1 | 6 | 610 | ||
GenBank mRNA | BC086352.1 | - | 6 | 610 | 7303026 | |
FQ220233.1 | - | 6 | 633 |