Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Csn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031562.1 | NP_113750.1 | 2 | 76 | ||
GenBank mRNA | K02598.1 | - | 2 | 76 |