Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr182 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053302.3 | NP_445754.2 | 2 | 524 | ||
XM_008765379.1 | XP_008763601.1 | 2 | 838 | |||
XM_008765380.1 | XP_008763602.1 | 1 | 888 | |||
GenBank mRNA | BC129072.1 | - | 2 | 524 | 7096756 | |
S79811.1 | - | 2 | 602 |
Set Membership: |
G-protein coupled receptor |