Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp511 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106309.1 | NP_001099779.1 | 3, 4 | 505 | ||
XM_008759974.1 | XP_008758196.1 | 3, 4 | 440 | |||
GenBank mRNA | FQ226365.1 | - | 3, 4 | 478 |