Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubl4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106346.2 | NP_001099816.1 | 4 | 506 | ||
GenBank mRNA | BC166513.1 | - | 4 | 506 | 7129152 |