Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc12a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019229.2 | NP_062102.1 | 13, 14 | 1745 | ||
XM_006255482.2 | XP_006255544.1 | 13, 14 | 3149 | |||
XM_008772492.1 | XP_008770714.1 | 12, 13 | 1736 | |||
GenBank mRNA | U55815.1 | - | 13, 14 | 1745 |