Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Btg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017258.1 | NP_058954.1 | 2 | 210 | ||
GenBank mRNA | L26268.1 | - | 2 | 210 |