Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uap1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134516.1 | NP_001127988.1 | 3, 4 | 751 | ||
GenBank mRNA | BC168670.1 | - | 3, 4 | 751 | 7935165 |