Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Edf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106557.1 | NP_001100027.1 | 5 | 482 | ||
GenBank mRNA | BC158849.1 | - | 5 | 437 | 7445841 |