Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dguok | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106602.1 | NP_001100072.1 | 3 | 371 | ||
XM_006236735.2 | XP_006236797.1 | 3 | 400 | |||
XM_006236736.2 | XP_006236798.1 | 3 | 404 | |||
XM_006236737.2 | XP_006236799.1 | 3 | 397 | |||
XM_008763056.1 | XP_008761278.1 | 3 | 404 | |||
XM_008763057.1 | XP_008761279.1 | 3 | 397 | |||
XM_008763058.1 | XP_008761280.1 | 3 | 161 | |||
GenBank mRNA | BC168743.1 | - | 3 | 312 | 6919887 |