Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klre1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181372.2 | NP_852037.1 | 1, 2 | 198 | ||
XM_006237081.2 | XP_006237143.1 | 3, 4 | 503 | |||
XM_006237082.2 | XP_006237144.1 | 2, 3 | 585 | |||
XM_008763311.1 | XP_008761533.1 | 2, 3 | 582 | |||
GenBank mRNA | AF486185.1 | - | 1, 2 | 139 | ||
AF486186.1 | - | 1, 2 | 198 | |||
AF486187.1 | - | 2, 3 | 286 | |||
FQ221012.1 | - | 2, 3 | 296 |