Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grhpr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113754.1 | NP_001107226.1 | 1, 2 | 150 | ||
GenBank mRNA | BC158680.1 | - | 1, 2 | 134 | 8373346 | |
FQ210631.1 | - | 1, 2 | 97 |