Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grhpr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113754.1 | NP_001107226.1 | 7 | 777 | ||
GenBank mRNA | BC158680.1 | - | 7 | 761 | 8373346 | |
FQ210631.1 | - | 7 | 724 |