Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC299282 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182474.1 | NP_872280.1 | 3, 4 | 677 | ||
GenBank mRNA | AY298742.1 | - | 3, 4 | 677 | ||
AY325133.1 | - | 3, 4 | 677 | |||
BC088096.1 | - | 3, 4 | 1001 | 7372979 | ||
D00752.1 | - | 3, 4 | 984 | |||
FQ209483.1 | - | 3, 4 | 997 | |||
FQ210703.1 | - | 3, 4 | 999 | |||
FQ211200.1 | - | 1, 2 | 72 | |||
FQ218472.1 | - | 3, 4 | 1003 | |||
M15917.1 | - | 2, 3 | 701 | |||
X16357.1 | - | 3, 4 | 997 |