Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mmp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134530.1 | NP_001128002.1 | 5 | 706 | ||
GenBank mRNA | BC169019.1 | - | 5 | 706 | 9091657 |