Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lysmd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106839.1 | NP_001100309.1 | 2 | 451 | ||
XM_006243405.2 | XP_006243467.1 | 2 | 1344 | |||
XM_006243406.2 | XP_006243468.1 | 3 | 1829 | |||
XM_008766248.1 | XP_008764470.1 | 3 | 311 | |||
GenBank mRNA | BC167017.1 | - | 2 | 343 | 7302522 |