Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC301124 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134533.1 | NP_001128005.1 | 4 | 511 | ||
XM_006244315.2 | XP_006244377.1 | 4 | 1478 |