Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mageb5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106946.1 | NP_001100416.1 | 2 | 554 | ||
XM_006257012.2 | XP_006257074.1 | 2 | 528 | |||
XM_006257013.2 | XP_006257075.1 | 2 | 532 |