Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tssk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012019.1 | NP_001012019.1 | 1 | 316 | ||
GenBank mRNA | BC079037.1 | - | 1 | 316 | 7114351 |