Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100910838 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751152.3 | XP_003751200.2 | 4 | 2585 | ||
XM_006249084.2 | XP_006249146.1 | 3 | 2944 | |||
XM_006249085.2 | XP_006249147.1 | 4 | 2254 | |||
XM_006249087.2 | XP_006249149.1 | 4 | 1994 | |||
XM_006249089.2 | XP_006249151.1 | 3 | 2943 |
Gene Symbol: LOC103690980 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008758796.1 | XP_008757018.1 | 1 | 103 | ||
XM_008758797.1 | XP_008757019.1 | 1 | 103 |
Gene Symbol: Nyap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107132.1 | NP_001100602.1 | 4 | 2393 | ||
XM_006249143.2 | XP_006249205.1 | 2943 | ||||
XM_006249144.2 | XP_006249206.1 | 4 | 2585 | |||
XM_006249145.2 | XP_006249207.1 | 4 | 2253 | |||
XM_006249147.2 | XP_006249209.1 | 4 | 1993 | |||
XM_006249149.2 | XP_006249211.1 | 2943 |