Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apon | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009385.1 | NP_001009385.1 | 2 | 175 | ||
GenBank mRNA | BC088168.1 | - | 2 | 175 | 7322067 |