Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcg3l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107205.1 | NP_001100675.1 | 15, 16 | 1847 | ||
XM_006250606.2 | XP_006250668.1 | 15, 16 | 2486 |