Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1qtnf7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107221.1 | NP_001100691.1 | 3 | 876 | ||
XM_006251073.2 | XP_006251135.1 | 3 | 1434 | |||
XM_006251075.2 | XP_006251137.1 | 3 | 2137 | |||
XM_008770208.1 | XP_008768430.1 | 2 | 1374 | |||
GenBank mRNA | BC166720.1 | - | 3 | 1264 | 7932439 | |
FQ220553.1 | - | 3 | 874 |