Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp219 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007681.1 | NP_001007682.1 | 2 | 378 | ||
XM_006251878.2 | XP_006251940.1 | 3 | 1537 | |||
XM_006251879.2 | XP_006251941.1 | 4 | 1195 | |||
XM_006251880.2 | XP_006251942.1 | 3 | 921 | |||
XM_006251882.2 | XP_006251944.1 | 2 | 832 | |||
XM_008770522.1 | XP_008768744.1 | 3 | 1221 | |||
GenBank mRNA | BC082017.1 | - | 2 | 378 | 7132141 | |
FQ212172.1 | - | 3 | 361 | |||
FQ221971.1 | - | 4 | 785 |