Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ngdn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107263.1 | NP_001100733.1 | 5, 6 | 426 | ||
XM_006251966.2 | XP_006252028.1 | 5, 6 | 619 | |||
XM_008770676.1 | XP_008768898.1 | 5, 6 | 1411 |