Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Polr3d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031653.1 | NP_001026823.1 | 3 | 235 | ||
XM_006252272.2 | XP_006252334.1 | 3 | 235 | |||
GenBank mRNA | BC099167.1 | - | 3 | 235 | 7382125 |