Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC306079 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047879.1 | NP_001041344.1 | 1 | 66 | ||
GenBank mRNA | AY387052.1 | - | 1 | 66 |