Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem59l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271055.1 | NP_001257984.1 | 7, 8 | 936 | ||
XM_006252897.2 | XP_006252959.1 | 8, 9 | 1006 | |||
GenBank mRNA | BC089056.1 | - | 7, 8 | 863 | ||
FQ212329.1 | - | 7, 8 | 911 | |||
FQ214027.1 | - | 7, 8 | 891 |