Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Akr1c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033697.1 | NP_001028869.1 | 1, 2 | 87 | ||
XM_006254188.2 | XP_006254250.1 | 1, 2 | 141 | |||
GenBank mRNA | AB300410.1 | - | 1, 2 | 78 | ||
BC088227.1 | - | 1, 2 | 76 | |||
BC104716.1 | - | 1, 2 | 87 | 7373256 |