Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnfaip8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107387.1 | NP_001100857.1 | 4 | 975 | ||
XM_006254708.2 | XP_006254770.1 | 2 | 6887 | |||
XM_006254709.2 | XP_006254771.1 | 2 | 627 | |||
XM_008772149.1 | XP_008770371.1 | 2 | 2965 | |||
GenBank mRNA | FQ226088.1 | - | 2 | 645 | ||
FQ230976.1 | - | 2 | 645 | |||
FQ235239.1 | - | 2 | 645 |