Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptgdr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012070.1 | NP_001012070.1 | 1 | 112 | ||
XM_006231040.2 | XP_006231102.1 | 1 | 200 | |||
XM_008760231.1 | XP_008758453.1 | 2 | 178 | |||
GenBank mRNA | AY228550.1 | - | 1 | 112 |
Set Membership: |
G-protein coupled receptor |