Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdc14a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107718.2 | NP_001101188.2 | 1, 2 | 666 | ||
NM_001134856.1 | NP_001128328.1 | 1, 2 | 592 | |||
XM_008761460.1 | XP_008759682.1 | 1, 2 | 71 | |||
GenBank mRNA | BC161876.1 | - | 1, 2 | 666 | 7115002 |