Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1qtnf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107745.1 | NP_001101215.1 | 2 | 1075 | ||
GenBank mRNA | BC168928.1 | - | 2 | 1088 | 8368551 |