Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bai2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107914.1 | NP_001101384.1 | 25, 26 | 3744 | ||
XM_006238934.1 | XP_006238996.1 | 26, 27 | 3852 | |||
XM_006238935.2 | XP_006238997.1 | 26, 27 | 4215 | |||
XM_006238936.1 | XP_006238998.1 | 26, 27 | 3852 | |||
XM_006238937.1 | XP_006238999.1 | 25, 26 | 3753 | |||
XM_006238938.1 | XP_006239000.1 | 25, 26 | 3687 | |||
XM_006238939.1 | XP_006239001.1 | 25, 26 | 3687 | |||
XM_008764141.1 | XP_008762363.1 | 25, 26 | 3753 | |||
XM_008764142.1 | XP_008762364.1 | 24, 25 | 3588 | |||
XM_008764143.1 | XP_008762365.1 | 24, 25 | 3588 |
Set Membership: |
G-protein coupled receptor |