Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zc3h7b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130695.1 | NP_001124167.1 | 20, 21 | 2563 | ||
XM_006242085.2 | XP_006242147.1 | 21, 22 | 2847 | |||
XM_008765719.1 | XP_008763941.1 | 21, 22 | 3933 |