Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Copz1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108117.1 | NP_001101587.1 | 6 | 428 | ||
GenBank mRNA | FQ226451.1 | - | 6 | 404 | ||
FQ234671.1 | - | 6 | 404 |