Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp142 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108225.1 | NP_001101695.1 | 6 | 4061 | ||
XM_006245222.2 | XP_006245284.1 | 6 | 3252 | |||
XM_008767240.1 | XP_008765462.1 | 7 | 3689 | |||
XM_008767241.1 | XP_008765463.1 | 6 | 3289 | |||
XM_008767242.1 | XP_008765464.1 | 6 | 3019 | |||
XM_008767243.1 | XP_008765465.1 | 8 | 3853 | |||
XM_008767244.1 | XP_008765466.1 | 8 | 3846 | |||
XM_008767245.1 | XP_008765467.1 | 9 | 3679 | |||
XM_008767246.1 | XP_008765468.1 | 9 | 3648 | |||
XM_008767247.1 | XP_008765469.1 | 9 | 3623 | |||
XM_008767248.1 | XP_008765470.1 | 9 | 3592 | |||
GenBank mRNA | BC100153.1 | - | 7 | 3327 |