Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Morf4l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007714.1 | NP_001007715.1 | 4 | 497 | ||
XM_006257295.1 | XP_006257357.1 | 4 | 533 | |||
XM_006257296.2 | XP_006257358.1 | 4 | 527 | |||
XM_006257297.1 | XP_006257359.1 | 3 | 432 | |||
GenBank mRNA | AY539870.1 | - | 1 | 212 | ||
BC083606.1 | - | 4 | 497 | 7133330 | ||
FQ212552.1 | - | 3 | 395 | |||
FQ232538.1 | - | 4 | 496 |