Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Taar8a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175599.1 | NP_783189.1 | 1 | 49 |
Gene Symbol: LOC100910174 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003748672.3 | XP_003748720.1 | 1 | 41 |
Set Membership: |
G-protein coupled receptor |