Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zbtb16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013181.1 | NP_001013199.1 | 2 | 262 | ||
GenBank mRNA | AY781102.1 | - | 2 | 262 | ||
AY781103.1 | - | 2 | 262 | |||
AY781104.1 | - | 2 | 253 |