Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ska3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108379.1 | NP_001101849.1 | 4 | 888 | ||
XM_006252074.1 | XP_006252136.1 | 4 | 694 | |||
XM_006252075.1 | XP_006252137.1 | 4 | 694 | |||
XM_008770758.1 | XP_008768980.1 | 4 | 691 | |||
GenBank mRNA | BC166461.1 | - | 4 | 665 | 9030107 |