Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smim19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002728416.4 | XP_002728462.1 | 2 | 338 | ||
GenBank mRNA | FQ214349.1 | - | 2 | 309 |