Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elmo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108415.1 | NP_001101885.1 | 7, 8 | 939 | ||
XM_008771735.1 | XP_008769957.1 | 7, 8 | 5840 | |||
GenBank mRNA | HQ438489.1 | - | 6, 7 | 439 |