Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elmo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108415.1 | NP_001101885.1 | 17 | 1965 | ||
XM_006254087.2 | XP_006254149.1 | 2 | 10578 | |||
XM_008771735.1 | XP_008769957.1 | 17 | 6866 | |||
GenBank mRNA | HQ438489.1 | - | 17 | 1510 |