Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dxo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_212497.2 | NP_997662.1 | 5 | 1021 | ||
GenBank mRNA | BC082083.1 | - | 5 | 1021 | 7133445 |