Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: H2afy2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135807.1 | NP_001129279.1 | 2, 3 | 361 | ||
GenBank mRNA | BC161981.1 | - | 2, 3 | 361 | 9029999 |